missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 13 publications
Marque: R&D Systems AF2199
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Legumain/Asparaginyl Endopeptidase Polyclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Spécification
| Legumain/Asparaginyl Endopeptidase | |
| Polyclonal | |
| Unconjugated | |
| Q99538 | |
| LGMN | |
| Mouse myeloma cell line NS0-derived recombinant human Legumain/Asparaginyl Endopeptidase Ile18-Tyr433 Accession # Q99538 | |
| 100 μg | |
| Primary | |
| Detects human Legumain/Asparaginyl Endopeptidase in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5641 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu