missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lgr6 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems FAB84581P-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Lgr6 Monoclonal specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry.
Spécification
| Lgr6 | |
| Monoclonal | |
| PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
| NS0 mouse myeloma cell line transfected with human Lgr6, Met1-Val967, Accession # Q9HBX8 | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 918726 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9HBX8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 59352 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu