missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB2/CD85d/ILT4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marque: R&D Systems AF2078
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ILT4/CD85d/LILRB2 Polyclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
Spécification
| ILT4/CD85d/LILRB2 | |
| Polyclonal | |
| Unconjugated | |
| Q8N423 | |
| LILRB2 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4 Gly24-His458 Accession # Q8N423 | |
| 100 μg | |
| Primary | |
| Detects human LILRB2/CD85d/ILT4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human (rh) ILT2 is observed and 5% cross-reactivity with rhILT5 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 3-25 ug/mL, Neutralization 0.3-1.0 ug/mL, CyTOF-ready | |
| CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10288 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu