Learn More
Abnova™ Human LINS1 Full-length ORF (AAH10363.1, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00055180-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
The Drosophila segment polarity gene lin encodes a protein, lines, which plays important roles in development of the epidermis and hindgut. This gene encodes a protein containing a lines-like domain. This gene is located on chromosome 15 and clustered with the gene encoding ankyrin repeat and SOCS box-containing protein 7. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: MKVFCEVLEELYKKVLLGATLENDSHDYVFYLNPAVSDQDCSTATSLEWANTCGIQGRHQPISVGVAPIAVAPVCLKTNSQMSGSREVMLLQLTVIKVMTTRILSVKTEFHAKEQYRDVIKILLESAKVDSKLICMFQNSDKLLSHMAAQCLALLLYFQLREKITLSNSWIAFCQKNLSEYSESNKAIYCLWTLTAIIKEIFKDSCSQKTEILKQFLTHFDTIFEVFYNSLFSQHFENCRDTSKIVNILMCFLDLLELLIASRIHLKLHFTCQRILFLKPSCMLEVITWPIQAFVKRKVIIFLKKCLLCKVGEDLCRGSVPALMPPDHHVAVDMLALANAVLQAVNSGLLKTLSVYEKHSFFGGDEVQPECELITSPDHVILRAASLVIMKSLEIKFQNYSSASEMKGNSPNSFCMQCVIIYLSTVIHNYQISGLVSpécification
AAH10363.1 | |
Liquid | |
55180 | |
LINS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKVFCEVLEELYKKVLLGATLENDSHDYVFYLNPAVSDQDCSTATSLEWANTCGIQGRHQPISVGVAPIAVAPVCLKTNSQMSGSREVMLLQLTVIKVMTTRILSVKTEFHAKEQYRDVIKILLESAKVDSKLICMFQNSDKLLSHMAAQCLALLLYFQLREKITLSNSWIAFCQKNLSEYSESNKAIYCLWTLTAIIKEIFKDSCSQKTEILKQFLTHFDTIFEVFYNSLFSQHFENCRDTSKIVNILMCFLDLLELLIASRIHLKLHFTCQRILFLKPSCMLEVITWPIQAFVKRKVIIFLKKCLLCKVGEDLCRGSVPALMPPDHHVAVDMLALANAVLQAVNSGLLKTLSVYEKHSFFGGDEVQPECELITSPDHVILRAASLVIMKSLEIKFQNYSSASEMKGNSPNSFCMQCVIIYLSTVIHNYQISGLV | |
RUO | |
LINS1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10583/WINS1 | |
LINS1 | |
Yes | |
wheat germ expression system |