missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LOX-1/OLR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Marque: R&D Systems AF1798
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LOX-1/OLR1 Polyclonal specifically detects LOX-1/OLR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Spécification
| LOX-1/OLR1 | |
| Polyclonal | |
| Unconjugated | |
| P78380 | |
| OLR1 | |
| Mouse myeloma cell line NS0-derived recombinant human LOX-1 (R&D Systems, Catalog # 1798-LX) Ser61-Gln273 Accession # P78380 | |
| 100 μg | |
| Primary | |
| Selected for its ability to block receptor-ligand interaction in a functional ELISA assay. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse LOX-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
| CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4973 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu