missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lymphotoxin beta R/TNFRSF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00€ - 541.00€
Spécification
| Antigène | Lymphotoxin beta R/TNFRSF3 |
|---|---|
| Clone | 71315 |
| Dilution | Agonist Activity 1.5-9 ng/mL |
| Classification | Monoclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18738838
![]() |
R&D Systems
MAB6291-SP |
25 μg |
209.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18088821
![]() |
R&D Systems
MAB6291-100 |
100 μg |
541.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Beschreibung
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Agonist Activity.Spezifikation
| Lymphotoxin beta R/TNFRSF3 | |
| Agonist Activity 1.5-9 ng/mL | |
| Unconjugated | |
| RUO | |
| Human | |
| P36941 | |
| 4055 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 71315 | |
| Monoclonal | |
| Mouse | |
| Agonist Activity | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| Mouse myeloma cell line NS0-derived recombinant human Lymphotoxin beta R/TNFRSF3 Gln31-Met227 Accession # P36941 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Lymphotoxin beta R/TNFRSF3 in direct ELISAs. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts