missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CX3CR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Marque: R&D Systems AF5825
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CX3CR1 Polyclonal specifically detects CX3CR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spécification
| CX3CR1 | |
| Polyclonal | |
| Unconjugated | |
| Q9Z0D9 | |
| CX3CR1 | |
| E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9 | |
| 100 μg | |
| Primary | |
| Detects human and mouse CX3CR1 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1524 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction