missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse GSK-3 alpha/beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB25063
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Beschreibung
GSK-3 alpha/beta Monoclonal specifically detects GSK-3 alpha/beta in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Spezifikation
| GSK-3 alpha/beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| P49841 | |
| GSK3A | |
| E. coli-derived recombinant human GSK-3 beta Asp200-Phe305 Accession # P49841 | |
| 100 μg | |
| Primary | |
| Detects human GSK-3 beta in direct ELISAs and human GSK-3 alpha and GSK-3 beta in Western blots. Detects human and mouse GSK-3 alpha/beta in immunocytochemistry. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 833916 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK3 alpha, GSK-3 alpha, GSK3 beta, GSK3A, GSK3B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2931 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur