missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse NLRP3/NALP3 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems IC7578P
424.15 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NLRP3/NALP3 Monoclonal specifically detects NLRP3/NALP3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Spécification
| NLRP3/NALP3 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| AGTAVPRL, AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CIAS1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCAS, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 114548 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 768319 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q8R4B8 | |
| NLRP3 | |
| E. coli-derived recombinant mouse NLRP3/NALP3 Met1-Arg153 Accession # Q8R4B8 | |
| 100 Tests | |
| Primary | |
| Detects mouse NLRP3/NALP3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human NLRP3/NALP3 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu