missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GDF-8/Myostatin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Marca: R&D Systems AF788
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GDF-8/Myostatin Polyclonal specifically detects GDF-8/Myostatin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Especificaciones
| GDF-8/Myostatin | |
| Polyclonal | |
| Unconjugated | |
| O08689 | |
| MSTN | |
| E. coli-derived recombinant mouse GDF-8/Myostatin Asp268-Ser376 Accession # O08689 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat GDF-8/Myostatin in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.6-3 ug/mL | |
| GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2660 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido