missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat LMW-PTP/ACP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marque: R&D Systems AF5075-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LMW-PTP/ACP1 Polyclonal specifically detects LMW-PTP/ACP1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Spécification
| LMW-PTP/ACP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acid phosphatase 1, soluble, acid phosphatase of erythrocyte, ACP1, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMWPTP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 52 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P24666 | |
| ACP1 | |
| E. coli-derived recombinant human LMW-PTP/ACP1 Ala2-His158 Accession # P24666 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat LMW-PTP/ACP1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu