missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat p27/Kip1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Marque: R&D Systems MAB2256
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Spécification
| p27/Kip1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P46527 | |
| CDKN1B | |
| E. coli-derived recombinant human p27 Met1-Thr198 Accession # P46527 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat p27 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 225501 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDKN1B, CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, Kip1, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1027 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu