Learn More
Human/Mouse/Rat Ribosomal Protein S6/RPS6 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marque: R&D Systems AF5436
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ribosomal Protein S6/RPS6 Polyclonal specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Spécification
| Ribosomal Protein S6/RPS6 | |
| Polyclonal | |
| Unconjugated | |
| P62753 | |
| RPS6 | |
| E. coli-derived recombinant human Ribosomal Protein S6 Met1-Lys249 Accession # P62753 | |
| 100 μg | |
| Primary | |
| Detects human, mouse and rat Ribosomal Protein S6 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6194 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |