missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nectin-2/CD112 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Marque: R&D Systems AF2229
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Nectin-2/CD112 Polyclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Spécification
| Nectin-2/CD112 | |
| Polyclonal | |
| Unconjugated | |
| NP_002847 | |
| PVRL2 | |
| Mouse myeloma cell line NS0-derived recombinant human Nectin-2/CD112 isoform a Gln32-Leu360 Accession # NP_002847 | |
| 100 μg | |
| Primary | |
| Detects human Nectin-2/CD112 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human (rh) Nectin-1, rhNectin-3 and rhNectin-4 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 1-15 ug/mL | |
| CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HVEB, HVEBpoliovirus receptor-like 2, MPH, nectin 2, Nectin2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2, PRR2nectin-2, PVRL2, PVRR2poliovirus receptor related 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5819 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu