Learn More
ACROBiosystems Human Nectin-2 / CD112 Protein, Fc Tag
Marque: ACROBiosystems PV2-H5253-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Poliovirus receptor-related 2 (PVRL2) is also known as nectin-2 and CD112 (formerly herpesvirus entry mediator B, HVEB), which is a human plasma membrane glycoprotein. PVRL2 is one of the plasma membrane components of adherens junctions. PVRL2 also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in PVRL2 gene have been associated with differences in the severity of multiple sclerosis. Also, PVRL2 binds with low affinity to TIGIT. Furthermore, PVRL2 acts as a receptor for herpes simplex virus 1 (HHV-1) mutant Rid1, herpes simplex virus 1 (HHV-2) and pseudorabies virus (PRV).
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_002847.1 | |
1 mg | |
Gln 32 - Leu 360 | |
CD112,Nectin-2,PVRL2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
62.2 kDa | |
Human Nectin-2, Fc Tag (PV2-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2). | |
-20°C | |
Nectin-2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |