Learn More
ACROBiosystems Human Nectin-2 / CD112 Protein, Fc Tag
350.80€ - 2419.31€
Spécification
Numéro d’adhésion | NP_002847.1 |
---|---|
Poids moléculaire | 62.2 kDa |
Source | Human Nectin-2, Fc Tag (PV2-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2). |
Immunogène | Gln 32 - Leu 360 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15857567
|
ACROBiosystems
PV2-H5253-100UG |
100 μg |
350.80€
100µg |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867567
|
ACROBiosystems
PV2-H5253-1MG |
1 mg |
2419.31€
1mg |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Poliovirus receptor-related 2 (PVRL2) is also known as nectin-2 and CD112 (formerly herpesvirus entry mediator B, HVEB), which is a human plasma membrane glycoprotein. PVRL2 is one of the plasma membrane components of adherens junctions. PVRL2 also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in PVRL2 gene have been associated with differences in the severity of multiple sclerosis. Also, PVRL2 binds with low affinity to TIGIT. Furthermore, PVRL2 acts as a receptor for herpes simplex virus 1 (HHV-1) mutant Rid1, herpes simplex virus 1 (HHV-2) and pseudorabies virus (PRV).
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_002847.1 | |
Human Nectin-2, Fc Tag (PV2-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2). | |
-20°C | |
Nectin-2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
62.2 kDa | |
Gln 32 - Leu 360 | |
CD112,Nectin-2,PVRL2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |