missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NK1R Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB6687
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NK1R Monoclonal specifically detects NK1R in Human samples. It is validated for Western Blot.
Spécification
| NK1R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P25103 | |
| TACR1 | |
| HEK293 human embryonic kidney cell line transfected with human NK1R Accession # P25103 | |
| 100 μg | |
| Primary | |
| Detects human NK1R in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 694549 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPR, SPRsubstance-P receptor, TAC1R, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor, TACR1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6869 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu