missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp46/NCR1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Marque: R&D Systems AF1850
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NKp46/NCR1 Polyclonal specifically detects NKp46/NCR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Spécification
| NKp46/NCR1 | |
| Polyclonal | |
| Unconjugated | |
| AAH64806 | |
| NCR1 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp46/NCR1 Gln22-Asn254 Accession # AAH64806 | |
| 100 μg | |
| Primary | |
| Detects human NKp46 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse NKp46 (MAR-1) is observed and less than 1% cross-reactivity with recombinant human (rh) NKp30, rhNKp44, and rhNKp80 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9437 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu