Learn More
Human OX40 Ligand/TNFSF4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Marque: R&D Systems AF1054
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
OX40 Ligand/TNFSF4 Polyclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Western Blot, ELISA, Neutralization.
Spécification
| OX40 Ligand/TNFSF4 | |
| Polyclonal | |
| Unconjugated | |
| Q6FGS4 | |
| TNFSF4 | |
| Mouse myeloma cell line NS0-derived recombinant human OX40 Ligand/TNFSF4 Gln51-Leu183 Accession # Q6FGS4 | |
| 100 μg | |
| Primary | |
| Detects human OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse OX40 Ligand/TNFSF4 is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.015-0.09 ug/mL | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7292 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |