missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Selectin/CD62P Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Marque: R&D Systems BBA32
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Western Blot, Adhesion Blockade.
Spécification
| P-Selectin/CD62P | |
| Polyclonal | |
| Unconjugated | |
| CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
| Sheep | |
| Protein A or G purified | |
| RUO | |
| 6403 | |
| Reconstitute with sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Adhesion Blockade | |
| LYOPH | |
| Western Blot 1 ug/mL, Adhesion Blockade | |
| SELP | |
| Chinese hamster ovary cell line CHO-derived recombinant human P-Selectin | |
| 500 μg | |
| Primary | |
| Selected for its ability to inhibit the binding of U937 cells to immobilized rhP-Selectin. In Western blots, less than 1% cross-reactivity is observed with rhE-Selectin and rhL-Selectin. | |
| Human | |
| Serum |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu