missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Peptidase Inhibitor 16/PI16 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marque: R&D Systems AF4980
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Peptidase Inhibitor 16/PI16 Polyclonal specifically detects Peptidase Inhibitor 16/PI16 in Human samples. It is validated for Western Blot.
Spécification
| Peptidase Inhibitor 16/PI16 | |
| Polyclonal | |
| Unconjugated | |
| AAH35634 | |
| PI16 | |
| Mouse myeloma cell line NS0-derived recombinant human Peptidase Inhibitor 16/PI16 Leu28-Leu437 Accession # AAH35634 | |
| 100 μg | |
| Primary | |
| Detects human Peptidase Inhibitor 16/PI16 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CRISP9, CRISP-9, Cysteine-rich secretory protein 9, dJ90K10.5, DKFZp586B1817, MGC45378, microseminoprotein, beta-binding protein, MSMBBP, peptidase inhibitor 16, PI16, PI-16, protease inhibitor 16, PSP94-binding protein, PSPBP | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 221476 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu