Learn More
Abnova™ Human PGK2 Partial ORF (NP_620061, 268 a.a. - 339 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00005232-Q01L
Informations supplémentaires : Poids : 0.00010kg
Description
The PGK2 gene encodes a testis-specific form of phosphoglycerate kinase (EC 2.7.2.3), which catalyzes the reversible conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate during glycolysis, generating one molecule of ATP. See also PGK1 (MIM 311800), which is ubiquitously expressed in all somatic tissues and maps to chromosome Xq13.[supplied by OMIM]
Sequence: DIMAKAQKNGVRITFPVDFVTGDKFDENAQVGKATVASGISPGWMGLDCGPESNKNHAQVVAQARLIVWNGPSpécification
NP_620061 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DIMAKAQKNGVRITFPVDFVTGDKFDENAQVGKATVASGISPGWMGLDCGPESNKNHAQVVAQARLIVWNGP | |
RUO | |
PGK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5232 | |
PGK2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PGK-2/PGKB/PGKPS/dJ417L20.2 | |
PGK2 | |
Recombinant | |
wheat germ expression system |