Learn More
Abnova™ Human PRDM2 Partial ORF (NP_036363, 1606 a.a. - 1705 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This tumor suppressor gene is a member of a nuclear histone/protein methyltransferase superfamily. It encodes a zinc finger protein that can bind to retinoblastoma protein, estrogen receptor, and the TPA-responsive element (MTE) of the heme-oxygenase-1 gene. Although the functions of this protein have not been fully characterized, it may (1) play a role in transcriptional regulation during neuronal differentiation and pathogenesis of retinoblastoma, (2) act as a transcriptional activator of the heme-oxygenase-1 gene, and (3) be a specific effector of estrogen action. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Spécification
Spécification
Numéro d’adhésion | NP_036363 |
À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 7799 |
Poids moléculaire | 36.74kDa |
Nom | PRDM2 (Human) Recombinant Protein (Q01) |
Test du contrôle qualité | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantité | 10 μg |
Immunogène | KTSRSLHVRVQKSKAVLQSKSTLASKKRTDRFNIKSRERSGGPVTRSLQLAAAADLSENKREDGSAKQELKDFSYSLRLASRCSPPAAPYITRQYRKVKA |
Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.