missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Progranulin/PGRN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB24201-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| Progranulin/PGRN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin | |
| Mouse myeloma cell line NS0-derived recombinant human Progranulin/PGRN Met1-Leu593 Accession # P28799 | |
| 100 μg | |
| Primary | |
| Detects human Progranulin/PGRN in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 296605 | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| P28799 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2896 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
RUO
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu