missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human QRSL1 Full-length ORF (AAH06084.1, 1 a.a. - 528 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00055278-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: MLGRSLREVSAALKQGQITPTELCQKCLSLIKKTKFLNAYITVSEEVALKQAEESEKRYKNGQSLGDLDGIPIAVKDNFSTSGIETTCASNMLKGYIPPYNATVVQKLLDQGALLMGKTNLDEFAMGSGSTDGVFGPVKNPWSYSKQYREKRKQNPHSENEDSDWLITGGSSGGSAAAVSAFTCYAALGSDTGGSTRNPAAHCGLVGFKPSYGLVSRHGLIPLVNSMDVPGILTRCVDDAAIVLGALAGPDPRDSTTVHEPINKPFMLPSLADVSKLCIGIPKEYLVPELSSEVQSLWSKAADLFESEGAKVIEVSLPHTSYSIVCYHVLCTSEVASNMARFDGLQYGHRCDIDVSTEAMYAATRREGFNDVVRGRILSGNFFLLKENYENYFVKAQKVRRLIANDFVNAFNSGVDVLLTPTTLSEAVPYLEFIKEDNRTRSAQDDIFTQAVNMAGLPAVSIPVALSNQGLPIGLQFIGRAFCDQQLLTVAKWFEKQVQFPVIQLQELMDDCSAVLENEKLASVSLKQSpécification
AAH06084.1 | |
Liquid | |
55278 | |
QRSL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGRSLREVSAALKQGQITPTELCQKCLSLIKKTKFLNAYITVSEEVALKQAEESEKRYKNGQSLGDLDGIPIAVKDNFSTSGIETTCASNMLKGYIPPYNATVVQKLLDQGALLMGKTNLDEFAMGSGSTDGVFGPVKNPWSYSKQYREKRKQNPHSENEDSDWLITGGSSGGSAAAVSAFTCYAALGSDTGGSTRNPAAHCGLVGFKPSYGLVSRHGLIPLVNSMDVPGILTRCVDDAAIVLGALAGPDPRDSTTVHEPINKPFMLPSLADVSKLCIGIPKEYLVPELSSEVQSLWSKAADLFESEGAKVIEVSLPHTSYSIVCYHVLCTSEVASNMARFDGLQYGHRCDIDVSTEAMYAATRREGFNDVVRGRILSGNFFLLKENYENYFVKAQKVRRLIANDFVNAFNSGVDVLLTPTTLSEAVPYLEFIKEDNRTRSAQDDIFTQAVNMAGLPAVSIPVALSNQGLPIGLQFIGRAFCDQQLLTVAKWFEKQVQFPVIQLQELMDDCSAVLENEKLASVSLKQ | |
RUO | |
QRSL1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564C1278/FLJ10989/FLJ12189/FLJ13447/GatA | |
QRSL1 | |
Yes | |
wheat germ expression system |