missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB7975
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Siglec-8 Monoclonal specifically detects Siglec-8 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Spécification
| Siglec-8 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9NYZ4 | |
| SIGLEC8 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-8 Met17-Ala363 Accession # Q9NYZ4 | |
| 100 μg | |
| Primary | |
| Detects human Siglec-8 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-7, -9, or -14 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 837535 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD329 antigen, CDw329, MGC59785, SAF2, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec8, Siglec-8, SIGLEC8L, SIGLEC8Lsialic acid-binding Ig-like lectin 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27181 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu