missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SNAP91 Partial ORF (NP_055656, 156 a.a. - 255 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00009892-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: MRTMAPEKLLKSMPILQGQIDALLEFDVHPNELTNGVINAAFMLLFKDLIKLFACYNDGVINLLEKFFEMKKGQCKDALEIYKRFLTRMTRVSEFLKVAESpécification
NP_055656 | |
Liquid | |
9892 | |
SNAP91 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP180/CALM/DKFZp781O0519/KIAA0656 | |
SNAP91 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRTMAPEKLLKSMPILQGQIDALLEFDVHPNELTNGVINAAFMLLFKDLIKLFACYNDGVINLLEKFFEMKKGQCKDALEIYKRFLTRMTRVSEFLKVAE | |
RUO | |
SNAP91 | |
Wheat Germ (in vitro) | |
GST |