missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX9 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 48 publications
Marque: R&D Systems AF3075-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SOX9 Polyclonal specifically detects SOX9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Spécification
| SOX9 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6662 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P48436 | |
| SOX9 | |
| E. coli-derived recombinant human SOX9 Met1-Lys151 Accession # P48436 | |
| 25 μg | |
| Primary | |
| Detects human SOX9 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human SOX10 is observed. | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu