Learn More
Abnova™ Human SURF2 Full-length ORF (NP_059973.2, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00006835-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene shares a bidirectional promoter with SURF1, which is located on the opposite strand. The function of the encoded protein from this housekeeping gene has not yet been determined. [provided by RefSeq]
Sequence: MSELPGDVRAFLREHPSLRLQTDARKVRCILTGHELPCRLPELQVYTRGKKYQRLVRASPAFDYAEFEPHIVPSTKNPHQLFCKLTLRHINKCPEHVLRHTQGRRYQRALCKYEECQKQGVEYVPACLVHRRRRREDQMDGDGPRPREAFWEPTSSDEGGAASDDSMTDLYPPELFTRKDLGSTEDGDGTDDFLTDKEDEKAKPPREKATDEGRRETTVYRGLVQKRGKKQLGSLKKKFKSHHRKPKSFSSCKQPGSpécification
NP_059973.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SURF-2 | |
SURF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6835 | |
SURF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSELPGDVRAFLREHPSLRLQTDARKVRCILTGHELPCRLPELQVYTRGKKYQRLVRASPAFDYAEFEPHIVPSTKNPHQLFCKLTLRHINKCPEHVLRHTQGRRYQRALCKYEECQKQGVEYVPACLVHRRRRREDQMDGDGPRPREAFWEPTSSDEGGAASDDSMTDLYPPELFTRKDLGSTEDGDGTDDFLTDKEDEKAKPPREKATDEGRRETTVYRGLVQKRGKKQLGSLKKKFKSHHRKPKSFSSCKQPG | |
RUO | |
SURF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |