missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACE/ADAM17 Ectodomain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Marque: R&D Systems MAB9301-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready, Knockout Validated.
Spécification
| TACE/ADAM17 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells | |
| P78536 | |
| ADAM17 | |
| Insect ovarian cell line T. ni-derived recombinant human TACE/ADAM17 Pro18-Asn671 Accession # P78536 | |
| 100 μg | |
| Primary | |
| Detects the ectodomain of human TACE/ADAM17 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with the ectodomain of recombinant human ADAM8, 9, 15 and recombinant mouse ADAM10 is observed. | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF, KnockDown | |
| 111633 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6868 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu