Learn More
ACROBiosystems Human THSD1 Protein, His Tag
Marque: ACROBiosystems TH1-H52H5-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Thrombospondin type-1 domain-containing protein 1 (THSD1) is also known as Transmembrane molecule with thrombospondin module (TMTSP), which is a single-pass type I membrane protein and contains a type 1 thrombospondin domain that is found in thrombospondin, a number of proteins involved in the complement pathway, as well as extracellular matrix proteins. In addition, THSD1 is widely expressed on endothelial cells, with highest expression in the lung. THSD1's ligand and function are unknown, but it is postulated that THSD1 may be involved in the regulation of vasculogenesis and/or angiogenesis through its interaction with its specific ligand.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 57-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH63842 | |
1 mg | |
Glu 25 - Ile 361 | |
THSD1,TMTSP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.7 kDa | |
Human THSD1, His Tag (TH1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ile 361 (Accession # AAH63842). | |
-20°C | |
THSD1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |