missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tie-2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems FAB3131N-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tie-2 Monoclonal antibody specifically detects Tie-2 in Human samples. It is validated for Immunophenotyping,Flow Cytometry
Spécification
| Tie-2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| Mouse myeloma cell line NS0-derived recombinant human Tie-2, Ala23-Lys745, Accession # AAA61139 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 83715 | |
| Flow Cytometry 5 uL/10^6 cells | |
| AAA61139 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7010 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu