missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TJAP1 Partial ORF (NP_542171.1, 77 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00093643-Q01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16151277

  • 359.00€ / 10µg
Expédition estimée: 13-07-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a tight junction-associated protein. Incorporation of the encoded protein into tight junctions occurs at a late stage of formation of the junctions. The encoded protein localizes to the Golgi and may function in vesicle trafficking. Alternatively spliced transcript variants have been described. A related pseudogene exists on the X chromosome. [provided by RefSeq]

Sequence: LELELGQSREELDKFKDKFRRLQNSYTASQRTNQELEDKLHTLIKKAEMDRKTLDWEIVELTNKLLDAKNTINKLEELNERYRLDCNPAVQLLKCNKSHFRNHK
Spécification

Spécification

NP_542171.1
Liquid
93643
TJAP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686F06131/PILT/TJP4
TJAP1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LELELGQSREELDKFKDKFRRLQNSYTASQRTNQELEDKLHTLIKKAEMDRKTLDWEIVELTNKLLDAKNTINKLEELNERYRLDCNPAVQLLKCNKSHFRNHK
RUO
TJAP1
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents
Certificats
Promotions

Promotions