Learn More
ACROBiosystems Human Transferrin R / CD71 Protein, His Tag
Marque: ACROBiosystems CD1-H5243-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 77.0 kDa. The protein migrates as 80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_003225.2 | |
1 mg | |
Cys 89 - Phe 760 | |
CD71,TFRC,TFR,sTfR,TfR1,Trfr,T9,p90 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
77.0 kDa | |
Human Transferrin R, His Tag (CD1-H5243) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1). | |
-20°C | |
Transferrin R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |