Learn More
Human Transglutaminase 3/TGM3 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Marque:  R&D Systems AF4604
Code nomenclature Nacres: NA.46
Code produit. 18012122
  • 560.00€ / 100µg
Description
Transglutaminase 3/TGM3 Polyclonal specifically detects Transglutaminase 3/TGM3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Transglutaminase 3/TGM3
Polyclonal
Unconjugated
NP_003236
TGM3
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Transglutaminase 3/TGM3, Ala2-Glu693, Accession # NP_003236
100 μg
Primary
Detects human Transglutaminase 3/TGM3 in direct ELISAs and Western blots.
Human
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
EC 2.3.2.13, MGC126250, protein-glutamine gamma-glutamyltransferase E, TG(E), TG3, TGase E, TGase-3, TgaseE, TGEMGC126249, TGM3, transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase E, transglutaminase-3
Sheep
Affinity Purified
RUO
7053
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.