missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM39 Partial ORF (NP_067076, 421 a.a. - 517 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00056658-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16152243

  • 508.00€ / 25µg
Expédition estimée: 05-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: TPLPETGYWRVRLWNGDKYAATTTPFTPLHIKVKPKRVGIFLDYEAGTLSFYNVTDRSHIYTFTDTFTEKLWPLFYPGIRAGRKNAAPLTIRPPTDW
Spécification

Spécification

NP_067076
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TPLPETGYWRVRLWNGDKYAATTTPFTPLHIKVKPKRVGIFLDYEAGTLSFYNVTDRSHIYTFTDTFTEKLWPLFYPGIRAGRKNAAPLTIRPPTDW
RUO
TRIM39
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56658
TRIM39 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC32984/RNF23/TFP/TRIM39B
TRIM39
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions