missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSG101 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB114381-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TSG101 Monoclonal antibody specifically detects TSG101 in Human samples. It is validated for Western Blot
Spécification
| TSG101 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
| TSG101 containing synthetic peptide, Accession # Q99816 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 1065916 | |
| Western Blot 2 μg/mL | |
| Q99816 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7251 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu