Learn More
Abnova™ Human WARS2 Full-length ORF (NP_957715.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010352-P01.10ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.00010kg
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQSILDMTAVLLACGINPEKSILFQQSQVSEHTQLSWILSCMVRLPRLQHLHQWKAKTTKQKHDGTVGLLTYPVLQAADILLYKSTHVPVGEDQVQHMELVQDLAQGFNKKYGEFFPVPESILSMCVLVFLTSpécification
NP_957715.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TrpRS | |
WARS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10352 | |
WARS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQSILDMTAVLLACGINPEKSILFQQSQVSEHTQLSWILSCMVRLPRLQHLHQWKAKTTKQKHDGTVGLLTYPVLQAADILLYKSTHVPVGEDQVQHMELVQDLAQGFNKKYGEFFPVPESILSMCVLVFLT | |
RUO | |
WARS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |