missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1/CD54 Antibody (1H4), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB110-58732APCCY7
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human samples. It is validated for Flow Cytometry.
Spécification
| ICAM-1/CD54 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ICAM1 | |
| Raji cells and spleen cells fused with NS1 cells. | |
| 0.1 mL | |
| Primary | |
| This antibody (clone 1H4) reacts with CD54 (ICAM-1), a 85-110 kDa type I transmembrane glycoprotein (receptor for rhinovirus) expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells; the expression of CD54 is upregulated by activation. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 1H4 | |
| Flow Cytometry | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
| 3383.0 | |
| Human | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu