missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDNK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24831-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The function of this protein remains unknown.Spécification
IDNK | |
Polyclonal | |
Unconjugated | |
IDNK | |
C9orf103, Gluconate kinase, IDNK, Probable gluconokinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
414328, 75731 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5T6J7, Q8R0J8 | |
IDNK | |
IDNK Fusion Protein Ag20088 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |