Learn More
Invitrogen™ IFN omega Human Matched Antibody Pair
Matched Antibody Pair
Marque: Invitrogen™ BMS233MST
Includes:
- 1 vial (1.1mL) monoclonal Coating Antibody to human IFN-omega (100μg/mL)
- 1 vial (55μL) Biotin-Conjugate anti-human IFN-omega monoclonal antibody
- 1 vial (11μL) Streptavidin-HRP
- 1 vial (30μL) human IFN-omega Standard protein, 60ng/mL
- 2 vials (50mL) Sample Diluent
Beschreibung
The Human IFN-β Matched Antibody Pair Kit comes with pre-matched antibody pairs,standards,sample diluent and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IFN-β. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
The protein encoded by this gene is produced by macrophages and has antiviral activity.
Spezifikation
| IFN omega | |
| Monoclonal | |
| P05000 | |
| IFNW1 | |
| RUO | |
| 3467 | |
| -20°C |
| ELISA | |
| Biotin | |
| IFNWP2, Interferon alpha-II-1, Interferon omega-1 | |
| 10 x 96 Tests | |
| Primary | |
| Human | |
| ELISA Kit |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.