missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15732-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
HSPB11 (Heat shock protein beta-11), also known as IFT25, was originally identified as a small heat shock protein. Recently it has been identified as a component of IFT complex B. It has been demonstrated that IFT25 is not required for ciliary assembly but is required for proper Hedgehog signaling, which in mammals occurs within cilia. Cilia lacking IFT25 have defects in the signal-dependent transport of multiple Hedgehog components and fail to activate the pathway upon stimulation.Spécification
IFT25 | |
Polyclonal | |
Unconjugated | |
HSPB11 | |
C1orf41, Heat shock protein beta 11, HSPB11, HSPCO34, IFT25, ITF25, Placental protein 25, PP25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51668, 685284, 72938 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D6H2, Q9Y547 | |
HSPB11 | |
IFT25 Fusion Protein Ag8490 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |