missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-3 Antibody (RM0096-11B16), Alexa Fluor™ 532, Novus Biologicals™

Code produit 18277065 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18277065 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18277065 Supplier Novus Biologicals Supplier No. NBP122452AF532

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigène IGFBP-3
Applications Western Blot
Classification Monoclonal
Clone RM0096-11B16
Conjugué Alexa Fluor 532
Dilution Western Blot
Formule 50 mM sodium borate with 0.05% sodium azide
Alias de gène acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Symboles de gène(s) IGFBP3
Espèces hôtes Rat
Immunogène Mouse recombinant protein of IGFBP-3
Méthode de purification Protein A or G purified
Quantité 0.1 mL
Disciplines de recherche Apoptosis, Biologically Active Proteins, Prostate Cancer
Primaire ou secondaire Primary
Identification génétique (Entrez) 3486.0
Spécificité du test This antibody detects mouse IGFBP-3 in Western blotting. No cross activity to other mouse IGFBPs.
Espèces cibles Mouse
Contenu et stockage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.