Learn More
IgG1 Mouse, Unconjugated, Clone: MOPC-21, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody
Marque:  Stemcell Technologies 60070
Encompass_Preferred
Code produit. 17148831
  • 243.00€ / 500µg
Description
  • The MOPC-21 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Spécification
Spécification
IgG1
Monoclonal
Unconjugated
500 μg
IgG1 κ
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
MOPC-21
Mouse
Isotype Control (Primary)
Other
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
IgG1 Mouse, Unconjugated, Clone: MOPC-21, Isotype Control, STEMCELL Technologies