missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-99049-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IKK alpha Polyclonal antibody specifically detects IKK alpha in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spécification
| IKK alpha | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human IKK alpha. | |
| 50 μg | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase, Signal Transduction | |
| 1147 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu