missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-1 beta/IL-1F2 Antibody (gevokizumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28830-50ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-1 beta/IL-1F2 Monoclonal antibody specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| Unconjugated | |
| P01584 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| gevokizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| IL-1b | |
| 50 μg | |
| Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis | |
| 3553 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu