missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-1 beta Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marque:  PeproTech 500-P51-1MG

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 17885463

  • 1834.00€ / 1mg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

AA Sequence of recombinant protein: MVPIRQLHYR LRDEQQKSLV LSDPYELKAL HLNGQNINQQ VIFSMSFVQG EPSNDKIPVA LGLKGKNLYL SCVMKDGTPT LQLESVDPKQ YPKKKMEKRF VFNKIEVKSK VEFESAEFPN WYISTSQAEH KPVFLGNNSG QDIIDFTMES VSS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-1beta. Anti-Murine IL-1beta-specific antibody was purified by affinity chromatography employing an immobilized Murine IL-1beta matrix. Sandwich ELISA: To detect Murine IL-1beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine IL-1beta (500-P51Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-1beta. Western Blot: To detect Murine IL-1beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine IL-1beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-1 beta
Polyclonal
Unconjugated
IL1B
catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Rabbit
Antigen affinity chromatography
RUO
16176
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P10749
IL1B
E.coli-derived Recombinant Murine IL-1beta.
1 mg
Primary
Mouse
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.