missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-10 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marque:  PeproTech 500-P139-100UG

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 17884203

  • 275.00€ / 100µg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

AA Sequence of recombinant protein: SKGHSIKGDN NCTHFPVSQT HMLRELRAAF SQVKTFFQKK DQLDNIVLTD SLLQDFKGYL GCQALSEMIK FYLVEVMPQA ENHGPEIKEH LNSLGEKLKT LWIQLRRCHR FLPCENKSKA VEQVKNDFNK LQDKGVYKAM NEFDIFINCI EAYVTLKMKN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-10. Anti-Rat IL-10-specific antibody was purified by affinity chromatography employing an immobilized Rat IL-10 matrix. Sandwich ELISA: To detect Rat IL-10 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat IL-10 (500-P139Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-10. Western Blot: To detect Rat IL-10 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat IL-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-10
Polyclonal
Unconjugated
IL10
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Rabbit
Antigen affinity chromatography
RUO
25325
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P29456
IL10
E.coli-derived Recombinant Rat IL-10.
100 μg
Primary
Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.