missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-17/IL-17A Antibody (SY18-VHH-11) - VHH-Fc, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28140-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| IL-17/IL-17A | |
| Monoclonal | |
| Unconjugated | |
| Q16552 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| SY18-VHH-11 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| CTLA-8/ IL-17a | |
| 100 μg | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| 3605 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu