Learn More
IL-33 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28035-1-AP-20UL
Code produit. 16845965
  • 140.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
The IL-33 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IL-33. This antibody recognizes mouse, rat antigen. The IL-33 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
IL-33
Polyclonal
Unconjugated
20 μL
77125
IgG
Western Blot, ELISA
200 μg/mL
Rabbit
Primary
Mouse, Rat